EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Multicomponent reaction of chalcones, malononitrile and DMF leading to γ-ketoamides.
PMID 25008794
Abstract
An efficient synthesis of γ-ketoamides was developed by the one-pot multicomponent reaction of chalcones, malononitrile and DMF (as both the reactant and solvent) in the presence of NaOH (3.0 equiv.). The reaction features high atom economy, easily available starting materials, operational simplicity, and good tolerance with diverse functional groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1407
Malononitrile, ≥99%
C3H2N2