EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Microbiology
Classification algorithm for subspecies identification within the Mycobacterium abscessus species, based on matrix-assisted laser desorption ionization-time of flight mass spectrometry.
PMID 25009048