EMAIL THIS PAGE TO A FRIEND
Oncology letters
A1BG and C3 are overexpressed in patients with cervical intraepithelial neoplasia III.
PMID 25009667
Abstract
The present study aimed to analyze sera proteins in females with cervical intraepithelial neoplasia, grade III (CIN III) and in healthy control females, in order to identify a potential biomarker which detects lesions that have a greater probability of cervical transformation. The present study investigated five sera samples from females who were Human Papilloma Virus (HPV) 16