EMAIL THIS PAGE TO A FRIEND
European review for medical and pharmacological sciences
Expression features of CXCR5 and its ligand, CXCL13 associated with poor prognosis of advanced colorectal cancer.
PMID 25010623
Abstract
CXCL13 plays a unique role in the trafficking and homing of B1 cells associated with its cognate receptor, CXCR5. The CXCR5-CXCL13 axis has been previously demonstrated to be a poor prognosis factor in malignancies. However, the clinical significance of the CXCR5-CXCL13 expression in colorectal cancer carcinoma (CRC) remains unclear. The aim of this study was to investigate the CXCR5-CXCL13 expression in CRC and determine its correlation with the progression and prognosis of the tumor. A total of 144 paraffin-embedded specimens with advanced colon cancer were assessed for CXCR5 and CXCL13 by immunohistochemistry. Patients' long-term survival was also monitored. There were significant differences in lymph node metastasis (p = 0.0066), neural invasion (p = 0.0061) and neural invasion (p = 0.0001) between high and low expression of CXCR5. There were significant differences in distant metastasis (p = 0.0261), TNM stage (p = 0.0409), differentiation (p < 0.0001) and neural invasion of the CXCL13. Both CXCR5 and CXCL13 was associated with poor correlation with the overall survival (OS) and relapse-free survival (RFS). Our data suggest that the CXCR5 and CXCL13 may play a crucial role in the development, metastasis and relapse of advanced colon cancer. They can be used as prognostic markers of colon cancer in clinical practice.