EMAIL THIS PAGE TO A FRIEND
Nanomedicine : nanotechnology, biology, and medicine
Effect of lipidated gonadotropin-releasing hormone peptides on receptor mediated binding and uptake into prostate cancer cells in vitro.
PMID 25014892
Abstract
Gonadotropin-releasing hormone (GnRH) receptors are overexpressed on many cancer cells but not on primary cell lines. This study was designed to investigate the targeting ability and uptake of dendritic lipidated [Gln(1)]-GnRH peptide analogues on receptor-positive prostate cancer PC-3 cells relative to receptor-negative ovarian carcinoma SKOV-3 cells for potential application in drug delivery. Direct antiproliferative effect of these was investigated on three GnRH-receptor positive cancer cells, PC-3, LNCaP and DU145. A significant dose dependent growth inhibitory effect was produced in DU145 cells by 5 dendrimers giving an IC50 value of 22-35 μM. All compounds were non-toxic to the normal peripheral blood mononuclear cells. This study demonstrates the use of specific dendritic lapidated GnRH analogues in growth inhibition of GnRH receptor positive prostate cancer cell lines, suggesting potential future clinical use of this or similar strategies to address GnRH receptor positive cancer cells.