EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Novel supports in chiral stationary phase development for liquid chromatography. Preparation, characterization and application of ordered mesoporous silica particles.
PMID 25015243
Abstract
Recent advances in the development of new materials are having a major impact on analytical chemistry. For example, the unique properties of ordered mesoporous silicas (OMSs) have been shown to enhance the analytical performance of many existing techniques or allow new, exciting ones to be developed. Likewise, the introduction of organo-functional groups makes OMSs highly versatile and enables them to perform specialized tasks, such as the separation of chiral compounds. This review provides an overview with the most relevant achievements in the preparation of OMS particles functionalized with chiral selectors. In addition, some examples from the last fifteen years regarding the analytical applications of functionalized OMS for chiral separations by high-performance liquid chromatography, ultra-high pressure high-performance liquid chromatography and capillary electrochromatography have been reviewed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
806870 Aluminum doped silica, mesoporous MCM-48, <150 μm particle size, pore size 3 nm, Cubic pore morphology
805890
Silica, mesoporous, 0.5 μm particle size, pore size ~2 nm
SiO2
806587
Silica, mesoporous, 2 μm particle size, pore size ~2 nm
SiO2
806765
Silica, mesoporous, 3 μm particle size, pore size ~2 nm
SiO2
806811
Silica, mesoporous, 1 μm particle size, pore size ~2 nm
SiO2
806900
Silica, mesoporous, 2 μm particle size, pore size ~4 nm
SiO2
806951
Silica, mesoporous, 3 μm particle size, pore size ~4 nm
SiO2
806889
Silica, mesoporous, 1 μm particle size, pore size ~4 nm
SiO2
806803
Silica, mesoporous SBA-15, <150 μm particle size, pore size 4 nm, Hexagonal pore morphology
SiO2
806854
Silica, mesoporous SBA-15, <150 μm particle size, pore size 8 nm, Hexagonal pore morphology
SiO2
806862
Silica, mesoporous SBA-15, <150 μm particle size, pore size 6 nm, Hexagonal pore morphology
SiO2
806927
Silica, mesoporous SBA-16, <150 μm particle size, pore size 5 nm, Cubic pore morphology
SiO2
805467
Silica, mesoporous MCM-48, 15 μm particle size, pore size 3 nm, Cubic pore morphology
O2Si
808989
Silica, mesoporous, 0.5 μm particle size, pore size ~4 nm
O2Si
806919 Titanium doped silica, mesoporous SBA-15, <150 μm particle size, pore size 4 nm, Hexagonal pore morphology