EMAIL THIS PAGE TO A FRIEND
Journal of Biomaterials Applications
Cellulose acetate butyrate/poly(caprolactonetriol) blends: Miscibility, mechanical properties, and in vivo inflammatory response.
PMID 25015652