EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Comprehensive comparison of cultivable norovirus surrogates in response to different inactivation and disinfection treatments.
PMID 25015883
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0759
Sodium glycochenodeoxycholate, ≥97% (HPLC)
C26H42NNaO5