EMAIL THIS PAGE TO A FRIEND
Neurobiology of disease
Region specific mitochondrial impairment in mice with widespread overexpression of alpha-synuclein.
PMID 25016198
Abstract
Parkinson's disease (PD) is characterized by the progressive degeneration of nigrostriatal dopaminergic neurons leading to motor deficits. The mechanisms underlying the preferential vulnerability of nigrostriatal dopaminergic neurons in PD remain poorly understood. Recent evidence supports a role for mitochondrial dysfunction and increased oxidative stress in PD pathogenesis. Genetic and pathological studies also point to alpha-synuclein as a critical factor in both familial and sporadic forms of the disease; alpha-synuclein pathology affects mitochondrial function but is widespread in PD brain, raising the question of its role in the greater vulnerability of nigrostriatal neurons in PD. We have examined mitochondrial function and oxidative damage in mice overexpressing human wild type alpha-synuclein broadly throughout the nervous system under the Thy1 promoter (Thy1-aSyn mice) between 4 and 8months of age. Similar levels of alpha-synuclein accumulation in mitochondria were detected in the ventral midbrain, striatum and cortex of Thy1-aSyn mice. However, analysis of mitochondrial respiration using Seahorse XF analyzer showed defects in mitochondrial respiratory complexes I, II, IV and V specifically in the midbrain, and IV and V in the striatum, of Thy1-aSyn mice compared to wild type littermates; mitochondrial complex I activity assay by ELISA confirmed a 40% inhibition specifically in the ventral midbrain. Mitochondrial dysfunction can contribute to oxidative stress and we observed a 40% increase in 4-hydroxynenal and 2-fold increase in malondialdehyde levels, indicative of a high level of lipid peroxidation, specifically in the ventral midbrain of Thy1-aSyn mice. The levels of peroxiredoxin 2, a neuronal antioxidant enzyme that is involved in removal of H2O2 and other toxic peroxides were decreased in the midbrain whereas its oxidized form increased 4-fold, suggesting that antioxidant defenses were compromised in this region. In contrast, peroxiredoxin 2 increased in the striatum and cortex, which may contribute to their protection in the presence of high levels of alpha-synuclein. Thus, in mice over-expressing alpha-synuclein, mitochondrial dysfunction occurred preferentially in nigrostriatal dopaminergic neurons many months before striatal dopamine loss occurs at 14months of age. This may contribute to a higher level of oxidative stress that overwhelms antioxidant defense in these neurons, leading to their increased vulnerability in PD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
SAB1405848
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
PRS3627
Anti-LAMP-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L0668
Anti-LAMP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029100
Anti-LAMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701987
Anti-LAMP2 antibody produced in rabbit
SAB2702058
Anti-LAMP2 antibody produced in rabbit
SAB1406520
Anti-PRDX2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Z373508
Carestream® BioMax® light film
AP127P Goat Anti-Mouse IgG Antibody, Fc, HRP conjugate, clone, 0.8 mg/mL, Chemicon®
AP156P Goat Anti-Rabbit IgG Antibody, Fc, HRP conjugate, clone, 0.8 mg/mL, Chemicon®
WH0002597M1
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
WH0007001M1
Monoclonal Anti-PRDX2 antibody produced in mouse, clone 4E10-2D2, purified immunoglobulin, buffered aqueous solution
SAB1404456
Monoclonal Anti-PRDX2 antibody produced in mouse, clone S2, purified immunoglobulin, buffered aqueous solution
69947
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
C7H15NO4S