EMAIL THIS PAGE TO A FRIEND
Clinica Chimica Acta; International Journal of Clinical Chemistry
Decreased serum levels of free fatty acids are associated with breast cancer.
PMID 25016244
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9673
Arachidonic acid, from porcine liver, ≥99% (GC), liquid
C20H32O2
A3555
Arachidonic acid, from porcine liver, BioReagent, suitable for cell culture, ≥99% (GC)
C20H32O2
A3925
Arachidonic acid, from porcine liver, ≥85% (capillary GC), liquid
C20H32O2
10931
Arachidonic acid, >95.0% (GC)
C20H32O2
A3611
Arachidonic acid, from non-animal source, ≥98.5% (GC)
C20H32O2
H5149
Heneicosanoic acid, analytical standard
C21H42O2
L1012
Linoleic acid, liquid, BioReagent, suitable for cell culture
C18H32O2
L1268
Linoleic acid, ≥93% (GC)
C18H32O2
62230
Linoleic acid, analytical standard
C18H32O2
W338001
Linoleic acid, ≥95%
C18H32O2
62240
Linoleic acid, technical, 58-74% (GC)
C18H32O2
L2376
Linolenic acid, ≥99%
C18H30O2
62170
Linolenic acid, ~70% (GC)
C18H30O2
62160
Linolenic acid, analytical standard
C18H30O2
43051
Palmitic acid, certified reference material, TraceCERT®
C16H32O2
1492007
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
C16H32O2
76119
Palmitic acid, analytical standard
C16H32O2
76122
Palmitic acid, purum, ≥98.0% (GC)
C16H32O2
PHR1120
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C16H32O2
P0090000
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
C16H32O2
P9417
Palmitoleic acid, ≥98.5% (GC), liquid
C16H30O2
76169
Palmitoleic acid, analytical standard
C16H30O2