EMAIL THIS PAGE TO A FRIEND
Journal of biomedical nanotechnology
A new X-ray activated nanoparticle photosensitizer for cancer treatment.
PMID 25016650
Abstract
Photodynamic therapy (PDT) has attracted ever-growing attention as a promising modality in the treatment of cancer. However, due to the poor tissue penetration by light, PDT has rarely been applied for treating deep-seated tumors. This problem can be solved if photosensitizers are activated by X-rays, which are able to penetrate deeply into tissues. Previous attempts using X-rays to activate photosensitizers were not very successful, since the traditional PDT photosensitizers could not efficiently be activated with X-rays. Here we exploit copper-cysteamine complex (Cu-Cy) nanoparticles as a new type of photosensitizer activated by X-ray for cancer treatment. The newly invented Cu-Cy nanoparticles can be activated directly by X-rays to produce singlet oxygen. In vitro and in vivo study on human breast cancer cells (MCF-7) have shown significant cell destruction using Cu-Cy nanoparticles under X-ray activation. The Cu-Cy nanoparticles are a novel and potent X-ray activated photosensitizer which can enable PDT for deep cancer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF27350287
Copper, wire reel, 1000m, diameter 0.1mm, annealed, 99.9%
Cu
GF63655780
Copper, wire reel, 1000m, diameter 0.25mm, annealed, 99.9%
Cu
GF47171033
Copper, wire reel, 100m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF18312791
Copper, wire reel, 100m, diameter 0.02mm, hard, 99.99+%
Cu
GF89712333
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Cu
GF93417105
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Cu
GF64298829
Copper, wire reel, 100m, diameter 0.02mm, annealed, 99.99%
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
GF39045090
Copper, wire reel, 100m, diameter 0.075mm, annealed, 99.9%
Cu
GF71409089
Copper, wire reel, 100m, diameter 0.125mm, annealed, 99.9%
Cu
GF42860651
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF69188079
Copper, wire reel, 100m, diameter 0.25mm, annealed, 99.9%
Cu
GF98066054
Copper, wire reel, 100m, diameter 0.5mm, annealed, 99.9%
Cu
GF81594399
Copper, wire reel, 100m, diameter 0.5mm, annealed, silver plated, 99.9%
Cu
GF82471620
Copper, wire reel, 100m, diameter 1.0mm, annealed, 99.9%
Cu
GF83100619
Copper, wire reel, 10m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF80026770
Copper, wire reel, 10m, diameter 0.02mm, hard, 99.99+%
Cu
GF74431324
Copper, wire reel, 10m, diameter 0.05mm, as drawn, 99.99+%
Cu
GF81301609
Copper, wire reel, 10m, diameter 0.75mm, as drawn, 99.98+%
Cu
GF86675439
Copper, wire reel, 10m, diameter 0.01mm, hard, 99.99%
Cu
GF77356109
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Cu
GF56973257
Copper, wire reel, 2000m, diameter 0.25mm, annealed, 99.9%
Cu