EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Naringenin inhibits migration of bladder cancer cells through downregulation of AKT and MMP‑2.
PMID 25017119
Abstract
Bladder cancer is one of the causes of cancer‑related death and has a high mortality rate due to its metastatic ability. Naringenin, a bioactive compound predominantly found in citrus fruits, exhibits several cellular functions, including anti‑oxidant, ‑lipidemia and ‑cancer abilities. However, the effects of naringenin on bladder cancer cells are yet to be elucidated. The present study investigated the molecular mechanisms underlying the effects of naringenin on the migration of TSGH‑8301 bladder cancer cells. Treatment with naringenin at doses ranging between 0 and 300 µM over a period of 24 h was found to reduce cell viability. Furthermore, zymography and western blot analysis revealed that naringenin reduced the expression of matrix metalloproteinase (MMP)‑2 in a dose‑dependent manner, and repressed its activity. Naringenin also reduced TSGH‑8301 cell migration in a concentration‑dependent manner, as evidenced by wound healing and Transwell® assays. In addition, naringenin was found to inhibit AKT activity and block the nuclear translocation of nuclear factor κ‑light‑chain‑enhancer of activated B cells. In conclusion, the findings of the present study show that naringenin is capable of inhibiting bladder cancer cell migration through the downregulation of the AKT and MMP‑2 pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
SAB1305558 MONOCLONAL ANTI-ERK2 antibody produced in mouse, clone 814CT5.4.2, IgG fraction of antiserum, buffered aqueous solution
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
WH0005594M1
Monoclonal Anti-MAPK1 antibody produced in mouse, clone 1D1, purified immunoglobulin, buffered aqueous solution
PLA0236
Rabbit anti-ERK2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0006855M1
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution