EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
A new biotinylating system for DNA using biotin aminocaproyl hydrazide and glutaraldehyde.
PMID 2501763
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3770
(+)-Biotinamidohexanoic acid hydrazide, ≥90% (TLC), powder
C16H29N5O3S