EMAIL THIS PAGE TO A FRIEND
Veterinary Immunology and Immunopathology
Identification of T cell receptor signaling pathway proteins in a feline large granular lymphoma cell line by liquid chromatography tandem mass spectrometry.
PMID 25017708