EMAIL THIS PAGE TO A FRIEND
Free Radical Biology & Medicine
Monoamine oxidase inhibition prevents mitochondrial dysfunction and apoptosis in myoblasts from patients with collagen VI myopathies.
PMID 25017965