PloS one
Up-regulation of microRNA-145 associates with lymph node metastasis in colorectal cancer.
PMID 25019299
Abstract
Metastasis is the main cause of mortality in patients with solid tumours. Identifying the exact molecules associated with CRC metastasis may be crucial to understand the process, which might also be translated to the diagnosis and treatment of CRC. In this study, we investigate the association of microRNA expression patterns with the lymph node metastasis of colorectal cancer. Among these candidate miRNAs, the expression of miRNA-145 was significantly related to lymph node metastasis of CRC. Both in vitro and in vivo study demonstrated that up-regulation of miR-145 could improve the ability of migration and invasion of colorectal cancer cell, while no effect on proliferation was observed. The mechanism of this promotion is associated with the stabilization of Hsp-27, a protein which plays an important role in the promotion of metastasis. These results may be crucial to understanding CRC metastasis and may be translated to the diagnosis and treatment of CRC.