PloS one
Simple signaling molecules for inductive bone regenerative engineering.
PMID 25019622
Abstract
With greater than 500,000 orthopaedic procedures performed in the United States each year requiring a bone graft, the development of novel graft materials is necessary. We report that some porous polymer/ceramic composite scaffolds possess intrinsic osteoinductivity as shown through their capacity to induce in vivo host osteoid mineralization and in vitro stem cell osteogenesis making them attractive synthetic bone graft substitutes. It was discovered that certain low crystallinity ceramics partially dissociate into simple signaling molecules (i.e., calcium and phosphate ions) that induce stem cells to endogenously produce their own osteoinductive proteins. Review of the literature has uncovered a variety of simple signaling molecules (i.e., gases, ions, and redox reagents) capable of inducing other desirable stem cell differentiation through endogenous growth factor production. Inductive simple signaling molecules, which we have termed inducerons, represent a paradigm shift in the field of regenerative engineering where they can be utilized in place of recombinant protein growth factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379980
Ammonium phosphate dibasic, ≥99.99% trace metals basis
H9N2O4P
215996
Ammonium phosphate dibasic, ACS reagent, ≥98%
H9N2O4P
A5764
Ammonium phosphate dibasic, reagent grade, ≥98.0%
H9N2O4P
09839
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
H9N2O4P
30402
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
H9N2O4P
04211
Ammonium phosphate dibasic, puriss., meets analytical specification of NF, 99-102%
H9N2O4P
C4955
Calcium nitrate tetrahydrate, BioXtra, ≥99.0%
CaN2O6 · 4H2O
C1396
Calcium nitrate tetrahydrate, ≥99.0%
CaN2O6 · 4H2O
C2786
Calcium nitrate tetrahydrate, suitable for cell culture, suitable for plant cell culture
CaN2O6 · 4H2O
C1000000
Cetylpyridinium chloride, European Pharmacopoeia (EP) Reference Standard
C21H38ClN · H2O
C0732
Cetylpyridinium chloride, meets USP testing specifications
C21H38ClN · H2O
1104006
Cetylpyridinium chloride, United States Pharmacopeia (USP) Reference Standard
C21H38ClN · H2O
C9756
Cholecalciferol, ≥98% (HPLC)
C27H44O
C9774
Cholecalciferol, analytical standard
C27H44O
C1357
Cholecalciferol, meets USP testing specifications
C27H44O
1131009
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
C27H44O
47763
Cholecalciferol (D3), analytical standard
C27H44O
PHR1237
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
C27H44O
740292
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
C27H44O
V-025
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material
C27H44O