EMAIL THIS PAGE TO A FRIEND
Journal of Dairy Science
The role of sodium in the salty taste of permeate.
PMID 25022679
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103128
2-Methyl-3-heptanone, 99%
C8H16O
414565
Divinylbenzene, technical grade, 80%
C10H10
169099
Divinylbenzene, technical grade, 55%
C10H10
17814
Lactose, tested according to Ph.Eur.
C12H22O11
791776
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
CK2O3
P5833
Potassium carbonate, BioXtra, ≥99.0%
CK2O3
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
367877
Potassium carbonate, 99.995% trace metals basis
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3
347825
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
CK2O3
60108
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
CK2O3
12611
Potassium carbonate, meets analytical specification of Ph. Helv., puriss., anhydrous, granulated, 99-101% (calc. to the dried substance)
CK2O3
900501
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
CK2O3
900502
Potassium carbonate, anhydrous, free-flowing, −325 mesh, Redi-Dri, reagent grade, ≥98%
CK2O3
T7158
Tetrabutylammonium bisulfate, ~99%
C16H37NO4S
86853
Tetrabutylammonium bisulfate, for ion pair chromatography, ≥99.0%
C16H37NO4S
86868
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
C16H37NO4S
86875
Tetrabutylammonium bisulfate, purum, ≥97.0% (T)
C16H37NO4S
72453
Tetrabutylammonium bisulfate solution, ~55% in H2O
C16H37NO4S
98479
Tetrabutylammonium bisulfate solution, for ion pair chromatography, concentrate, ampule
C16H37NO4S
791784
Tetrabutylammonium hydrogensulfate, anhydrous, free-flowing, Redi-Dri, 97%
C16H37NO4S
155837
Tetrabutylammonium hydrogensulfate, 97%
C16H37NO4S