EMAIL THIS PAGE TO A FRIEND
Cytokine
Interleukin-35 gene therapy exacerbates experimental rheumatoid arthritis in mice.
PMID 25022966
Abstract
Interleukin (IL)-35 was initially described as an immunosuppressive cytokine specifically produced by CD4(+)FoxP3(+) regulatory T cells (Treg). Since Treg play a major role in autoimmunity control and protect from inflammation, we aimed at evaluating the role of IL-35 in collagen-induced arthritis (CIA), a mouse model of rheumatoid arthritis (RA), using a non-viral gene transfer strategy. The clinical and histological effect of IL-35 was assessed in mice with CIA receiving an injection of two distinct plasmids encoding IL-35 gene (pIGneo-mIL-35 or pORF-mIL-35) 3 and 18 days after CIA induction. Treg and Th17 were characterized by flow cytometry in the spleen and lymph nodes of treated mice. Our results showed that whatever the plasmid used, IL-35 gene transfer resulted in a statistically significant increase in clinical scores of CIA compared to results with empty plasmid. The underlying cellular mechanisms of this effect were shown to be related to an increased Th17/Treg ratio in the spleen of pORF-mIL-35 treated mice. In conclusion, we show an unexpected but clear exacerbating effect of IL-35 gene transfer in an autoimmune and inflammatory RA model, associated with a modification of the Th17/Treg balance. Altogether, these result shows that this cytokine can promote chronic inflammation.