EMAIL THIS PAGE TO A FRIEND
Clinical rheumatology
Dynamic changes in the numbers of different subsets of peripheral blood NK cells in patients with systemic lupus erythematosus following classic therapy.
PMID 25024095
Abstract
Imbalance of natural killer (NK) cells is associated with the development of systemic lupus erythematosus (SLE). However, little is known about the dynamic changes on NK cells following therapy. This study aimed at examining the impact of classic therapies on the numbers of different subsets of NK cells in new-onset SLE patients. The numbers of different subsets of peripheral blood NK cells in 24 new-onset SLE patients before, 4 and 12 weeks post the classic therapies, and 7 healthy controls were determined by flow cytometry. The potential correlation between the numbers of NK cells and the values of clinical measures was analyzed. In comparison with that before treatment, the numbers of NK, NKG2C+, and KIR2DL3+ NK cells were significantly increased while the numbers of NKp46+ and NKG2A + NK cells significantly decreased at 4 and/or 12 weeks post the treatment only in the drug well-responding patients, but not in those poor responders (P < 0.05 for all). The numbers of NKG2C + NK cells were correlated positively with the levels of serum C3 while the numbers of KIR2DL3+ NK cells were correlated negatively with the scores of SLEDAI in these patients at 4 weeks post the treatment. The classic therapies modulated the numbers of some subsets of NK cells in drug well-responding SLE patients. The changes in the numbers of some subsets of NK cells may serve as biomarkers for evaluating the therapeutic responses of SLE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA030636
Anti-JAG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101188
Anti-JAG2 antibody produced in rabbit, affinity isolated antibody
SAB1401224
Anti-KIR3DL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406063
Anti-KLRC1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410734
Anti-KLRC1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1303125
ANTI-KLRC2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401225
Anti-KLRC2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1406823
Anti-NCR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4503644
Anti-NCR1 antibody produced in rabbit, affinity isolated antibody
SAB2701214
Anti-NCR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501094
Anti-NCR3 antibody produced in rabbit, affinity isolated antibody
SAB4700269 Monoclonal Anti-CD16 antibody produced in mouse, clone LNK16, purified immunoglobulin, buffered aqueous solution
SAB4700274 Monoclonal Anti-CD16 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
SAB4700276 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-154, purified immunoglobulin, buffered aqueous solution
SAB4700743 Monoclonal Anti-CD314 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
SAB4700452
Monoclonal Anti-CD56-FITC antibody produced in mouse, clone MEM-188, purified immunoglobulin, buffered aqueous solution
F0301 Monoclonal Anti-CD56-FITC antibody produced in mouse, clone C5.9, purified immunoglobulin, buffered aqueous solution
SAB1409259 Monoclonal Anti-JAG2 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB1403998
Monoclonal Anti-JAG2 antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
SAB1404010
Monoclonal Anti-KLRC1, (N-terminal) antibody produced in mouse, clone 2C3, ascites fluid
WH0003714M3
Monoclonal Anti-JAG2 antibody produced in mouse, clone 4F10, purified immunoglobulin, buffered aqueous solution
SAB4700268 Monoclonal Anti-CD16 antibody produced in mouse, clone MEM-168, purified immunoglobulin, buffered aqueous solution
SAB1302700
ANTI-KIR2DS1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2107772
Anti-KLRK1 antibody produced in rabbit, affinity isolated antibody
SAB2107773
Anti-KLRK1 antibody produced in rabbit, affinity isolated antibody
SAB1412940
Anti-NCR3 antibody produced in rabbit, purified immunoglobulin