EMAIL THIS PAGE TO A FRIEND
Applied Biochemistry and Biotechnology
Enhancing lung cancer diagnosis: electrochemical simultaneous bianalyte immunosensing using carbon nanotubes-chitosan nanocomposite.
PMID 25024132