EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer'S Disease
Long-term cannabidiol treatment prevents the development of social recognition memory deficits in Alzheimer's disease transgenic mice.
PMID 25024347