EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Bioactive constituents of Cirsium japonicum var. australe.
PMID 25025240
Abstract
Cirsium japonicum var. australe, used as a folk medicine in Taiwan, has been employed traditionally in the treatment of diabetes and inflammatory symptoms. Bioactivity-guided fractionation of its ethanolic extract, utilizing centrifugal partition chromatography monitored by DPPH-TLC analysis, led to the isolation of three new acetylenic phenylacrylic acid esters (1-3) and two new polyacetylenes (4 and 5), together with seven known compounds (6-12). The structures of 1-5 were elucidated by spectroscopic methods including 1D and 2D NMR techniques. The absolute configurations of 4 and 7 were determined utilizing Mosher's method and ECD/CD experiments. The DPPH scavenging activity of the constituents isolated from the C. japonicum var. australe ethanolic extract was evaluated. The potential antidiabetic activity of some of the isolates was evaluated using in vitro cellular glucose uptake and oil red staining assays.