EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
Isoliquiritigenin, a flavonoid from licorice, blocks M2 macrophage polarization in colitis-associated tumorigenesis through downregulating PGE2 and IL-6.
PMID 25026504