EMAIL THIS PAGE TO A FRIEND
The international journal of occupational and environmental medicine
Respiratory diseases in agate grinding workers in Iran.
PMID 25027041
Abstract
Agate is a hard silica stone with bands of various colors, which is used in jewelry. The agate grinding workers are exposed to silica dust. To determine the prevalence of respiratory diseases in agate grinding workers and the associated factors. In this cross-sectional study, 170 agate grinding workers from Mashhad, northeastern Iran, were examined. Medical and occupational history for respiratory illnesses was taken using respiratory questionnaire of the national program of silicosis control, lung examination, spirometry and chest radiography. Chest x-rays were interpreted according to the International Labor Office (ILO) classification system, 2000. The mean±SD of age and work duration of the participants were 31.2±10.1 and 13±8.2 years, respectively. The prevalence of silicosis among agate workers was 12.9% (95% CI: 7.9%-18.0%); 18 workers had simple and 4 had complicated silicosis. There was a significant (p<0.05) relationship between contracting silicosis and exposure duration. 20 (11.7%) workers had symptoms consistent with chronic bronchitis and 8 (4.7%) showed asthma and asthma-like symptoms. The most frequent disorder observed in spirometry was the restrictive pattern (n=43, 30%). In the agate grinders, clinical and spirometry findings did not match with radiological findings. Agate grinding workers are at increased risk for respiratory diseases, specifically for silicosis and chronic bronchitis. The disease is related to silica dust exposure, poor ventilation and inappropriate personal protection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
421553
Silica
O2Si
643645
Silica, mesostructured, MCM-41 type (hexagonal)
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
O2Si