EMAIL THIS PAGE TO A FRIEND
MMWR. Morbidity and mortality weekly report
Pedestrian traffic deaths among residents, visitors, and homeless persons--Clark County, Nevada, 2008-2011.
PMID 25029111
Abstract
Motor vehicle collisions and crashes are a leading cause of death among Nevada residents aged 5-34 years, representing 14% of all injury deaths in that age group in 2010. During 2008-2011, a total of 173 pedestrian deaths from motor vehicle collisions occurred in Nevada, accounting for 16% of motor vehicle deaths in the state. Approximately 75% (2 million persons) of Nevada residents live in Clark County, which includes the city of Las Vegas. To analyze pedestrian traffic deaths in Clark County among residents, visitors, and homeless persons, the Southern Nevada Health District used coroner's office data and death certificate data for the period 2008-2011. The results indicated that the average annual pedestrian traffic death rates from motor vehicle collisions during this period were 1.4 per 100,000 population for residents, 1.1 for visitors, and 30.7 for homeless persons. Among the three groups, time of day, location of motor vehicle collisions, and pedestrian blood alcohol concentration (BAC) differed. Effective interventions to increase roadway safety, such as lowering speed limits in areas with greater pedestrian traffic, targeting interventions during hours when alcohol-impaired walking is more likely, and modifying roadway designs to increase protection of pedestrians, might decrease pedestrian deaths among all three groups.