EMAIL THIS PAGE TO A FRIEND
Neoplasma
SKLB-287, a novel oral multikinase inhibitor of EGFR and VEGFR2, exhibits potent antitumor activity in LoVo colorectal tumor model.
PMID 25030434
Abstract
Colorectal cancer (CRC) is the third common cancer and most of the chemotherapies of CRC currently used often suffer limited efficacy and large side effects. Targeted small-molecule by anti-tumor drugs are thought a promising strategy for improving the efficacy and reducing the side effects. In this investigation, we report a novel multikinase inhibitor, termed SKLB-287, which was discovered by us recently. SKLB-287 could efficiently inhibit the activation of endothelial growth factor receptor (EGFR) and vascular endothelial growth factor receptor 2 (VEGFR2). It displayed very good anti-proliferative activity against LoVo CRC cells and considerable antiangiogenic potency in transgenic zebrafish embryos. Oral administration of SKLB-287 resulted in dose-dependent suppression of tumor growth in LoVo xenograft mouse model. Immunohistochemistry was adopted to examine the in vivo anti-tumor mechanism of action of SKLB-287.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018530
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
GW22768 Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA030069
Anti-MAPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
C7240 Anti-phospho-c-Met (pTyr1230/pTyr1234/pTyr1235) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
C6990 Anti-phospho-c-Met (pTyr1349) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4504579
Anti-phospho-EGFR (pSer1026) antibody produced in rabbit, affinity isolated antibody
SAB4504382
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504580
Anti-phospho-EGFR (pSer1071) antibody produced in rabbit, affinity isolated antibody
SAB4504578
Anti-phospho-EGFR (pSer695) antibody produced in rabbit, affinity isolated antibody
SAB4503759
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4300151
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody