EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Sixteenth biannual report of the Cochrane Haematological Malignancies Group: focus on Non-Hodgkin's lymphoma.
PMID 25031306
Abstract
This sixteenth biannual report of the Cochrane Haematological Malignancies Group highlights recently published randomized controlled trials (RCTs) in the field of hemato-oncology, with special focus on non-Hodgkin's lymphoma. The report covers the publication period June 2012 to July 2013. Trials are selected regarding their methodology and implication for clinical practice. Studies were identified by electronic search of MEDLINE using a broad search filter that covers all topics in hemato-oncology combined with a highly sensitive search filter for randomized trials (Cochrane Handbook for Systematic Reviews of Interventions). Four RCTs are presented in detail, followed by two further RCTs of high importance in a short version. The report is finalized with an overview of new and updated Cochrane Reviews.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0400
Carmustine, ≥98%
C5H9Cl2N3O2
C6645
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
C9H13N3O5 · HCl