EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Phasic dopamine neuron activity elicits unique mesofrontal plasticity in adolescence.
PMID 25031392
Abstract
The mesofrontal dopaminergic circuit, which connects the midbrain motivation center to the cortical executive center, is engaged in control of motivated behaviors. In addition, deficiencies in this circuit are associated with adolescent-onset psychiatric disorders in humans. Developmental studies suggest that the mesofrontal circuit exhibits a protracted maturation through adolescence. However, whether the structure and function of this circuit are modifiable by activity in dopaminergic neurons during adolescence remains unknown. Using optogenetic stimulation and in vivo two-photon imaging in adolescent mice, we found that phasic, but not tonic, dopamine neuron activity induces the formation of mesofrontal axonal boutons. In contrast, in adult mice, the effect of phasic activity diminishes. Furthermore, our results showed that dopaminergic and glutamatergic transmission regulate this axonal plasticity in adolescence and inhibition of dopamine D2-type receptors restores this plasticity in adulthood. Finally, we found that phasic activation of dopamine neurons also induces greater changes in mesofrontal circuit activity and psychomotor response in adolescent mice than in adult mice. Together, our findings demonstrate that the structure and function of the mesofrontal circuit are modifiable by phasic activity in dopaminergic neurons during adolescence and suggest that the greater plasticity in adolescence may facilitate activity-dependent strengthening of dopaminergic input and improvement in behavioral control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB152
Anti-Tyrosine Hydroxylase Antibody, Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, IF, IH, IH(P), IP and WB.
F7799
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA018531
Anti-C-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100832
Anti-FOS (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100833
Anti-FOS (ab2) antibody produced in rabbit, affinity isolated antibody
AV100639
Anti-FOS antibody produced in rabbit, IgG fraction of antiserum
SAB4500995
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB2103892
Anti-TH, (N-terminal) antibody produced in rabbit, affinity isolated antibody
T8700 Anti-Tyrosine Hydroxylase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots
D0731000 Dextran, European Pharmacopoeia (EP) Reference Standard
D0731005 Dextran 1, European Pharmacopoeia (EP) Reference Standard
D0734000 Dextran 10 for calibration, European Pharmacopoeia (EP) Reference Standard
D0737000 Dextran 250 for calibration, European Pharmacopoeia (EP) Reference Standard
D0733000 Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
D0735000 Dextran 40 for calibration, European Pharmacopoeia (EP) Reference Standard
D0738000 Dextran 40 for performance test, European Pharmacopoeia (EP) Reference Standard
D0739000 Dextran 60/70 for performance test, European Pharmacopoeia (EP) Reference Standard
D0736000 Dextran 70 for calibration, European Pharmacopoeia (EP) Reference Standard
D0732000 Dextran Vo, European Pharmacopoeia (EP) Reference Standard