EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Progressive disorganization of paranodal junctions and compact myelin due to loss of DCC expression by oligodendrocytes.
PMID 25031414
Abstract
Paranodal axoglial junctions are critical for maintaining the segregation of axonal domains along myelinated axons; however, the proteins required to organize and maintain this structure are not fully understood. Netrin-1 and its receptor Deleted in Colorectal Cancer (DCC) are proteins enriched at paranodes that are expressed by neurons and oligodendrocytes. To identify the specific function of DCC expressed by oligodendrocytes in vivo, we selectively eliminated DCC from mature myelinating oligodendrocytes using an inducible cre regulated by the proteolipid protein promoter. We demonstrate that DCC deletion results in progressive disruption of the organization of axonal domains, myelin ultrastructure, and myelin protein composition. Conditional DCC knock-out mice develop balance and coordination deficits and exhibit decreased conduction velocity. We conclude that DCC expression by oligodendrocytes is required for the maintenance and stability of myelin in vivo, which is essential for proper signal conduction in the CNS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405476
Anti-ABCC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA009433
Anti-CANX antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009696
Anti-CANX antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200084
Anti-CANX antibody produced in rabbit, whole antiserum
SAB5200097 Anti-CANX antibody produced in rabbit, affinity isolated antibody
SAB1405637
Anti-CNP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701826
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701825
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA040067
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300645
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
SAB2103104
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody, lyophilized powder
MAB386
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
HPA023138
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA022845
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
AB1987
Anti-Neurofilament M (145 kDa) Antibody, CT, serum, clone, Chemicon®
HPA008832
Anti-NFASC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32753
Anti-OLIG2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV31464
Anti-OLIG2 (AB2) antibody produced in rabbit, affinity isolated antibody
AV32752
Anti-OLIG2 (AB3) antibody produced in rabbit, affinity isolated antibody
HPA003254
Anti-OLIG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000368M1
Monoclonal Anti-ABCC6 antibody produced in mouse, clone 1E6, purified immunoglobulin, buffered aqueous solution
SAB1403563
Monoclonal Anti-ABCC6 antibody produced in mouse, clone 5D8, purified immunoglobulin, buffered aqueous solution
WH0000324M1
Monoclonal Anti-APC antibody produced in mouse, clone 3E2, purified immunoglobulin, buffered aqueous solution
SAB1402730
Monoclonal Anti-CNP antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution
PLA0125
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.