EMAIL THIS PAGE TO A FRIEND
Bioscience, Biotechnology, and Biochemistry
Lactate production as representative of the fermentation potential of Corynebacterium glutamicum 2262 in a one-step process.
PMID 25036691