PLoS ONE
Combined vildagliptin and metformin exert better cardioprotection than monotherapy against ischemia-reperfusion injury in obese-insulin resistant rats.
PMID 25036861
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8877
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
C19H15ClN4