EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Activation of a TRP-like channel and intracellular Ca2+ dynamics during phospholipase-C-mediated cell death.
PMID 25037570
Abstract
The model organism Neurospora crassa undergoes programmed cell death when exposed to staurosporine. Here, we show that staurosporine causes defined changes in cytosolic free Ca(2+) ([Ca(2+)]c) dynamics and a distinct Ca(2+) signature that involves Ca(2+) influx from the external medium and internal Ca(2+) stores. We investigated the molecular basis of this Ca(2+) response by using [Ca(2+)]c measurements combined with pharmacological and genetic approaches. Phospholipase C was identified as a pivotal player during cell death, because modulation of the phospholipase C signaling pathway and deletion of PLC-2, which we show to be involved in hyphal development, results in an inability to trigger the characteristic staurosporine-induced Ca(2+) signature. Using Δcch-1, Δfig-1 and Δyvc-1 mutants and a range of inhibitors, we show that extracellular Ca(2+) entry does not occur through the hitherto described high- and low-affinity Ca(2+) uptake systems, but through the opening of plasma membrane channels with properties resembling the transient receptor potential (TRP) family. Partial blockage of the response to staurosporine after inhibition of a putative inositol-1,4,5-trisphosphate (IP3) receptor suggests that Ca(2+) release from internal stores following IP3 formation combines with the extracellular Ca(2+) influx.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9754
2-Aminoethyl diphenylborinate, 97%
C14H16BNO
S4887
D-(+)-Sorbose, ≥99%
C6H12O6
X2628
Xestospongin C, film
C28H50N2O2