EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
Inhibition of p38 mitogen-activated protein kinase activation in the rostral anterior cingulate cortex attenuates pain-related negative emotion in rats.
PMID 25038392
Abstract
The emotional components of pain are far less studied than the sensory components. Previous studies have indicated that the rostral anterior cingulate cortex (rACC) is implicated in the affective response to noxious stimuli. Activation of p38 mitogen-activated protein kinase (MAPK) in the spinal cord has been documented to play an important role in diverse kinds of pathological pain states. We used formalin-induced conditioned place aversion (F-CPA) in rats, an animal model believed to reflect the emotional response to pain, to investigate the involvement of p38 MAPK in the rACC after the induction of affective pain. Intraplantar formalin injection produced a significant activation of p38 MAPK, as well as mitogen-activated kinase kinase (MKK) 3 and MKK6, its upstream activators, in the bilateral rACC. p38 MAPK was elevated in both NeuN-positive neurons and Iba1-positive microglia in the rACC, but not GFAP-positive cells. Blocking p38 MAPK activation in the bilateral rACC using its specific inhibitor SB203580 or SB239063 dose-dependently suppressed the formation of F-CPA. Inhibiting p38 MAPK activation did not affect formalin-induced two-phase spontaneous nociceptive response and low intensity electric foot-shock induced CPA. The present study demonstrated that p38 MAPK signaling pathway in the rACC contributes to pain-related negative emotion. Thus, a new pharmacological strategy targeted at the p38 MAPK cascade may be useful in treating pain-related emotional disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4502410
Anti-MAP2K3 antibody produced in rabbit, affinity isolated antibody
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
SAB4502409
Anti-MKK3 antibody produced in rabbit, affinity isolated antibody
SAB3701099 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4300119
Anti-phospho-MAP2K3 (pSer189) antibody produced in rabbit, affinity isolated antibody
SAB4504473
Anti-phospho-MKK3 (pSer189) antibody produced in rabbit, affinity isolated antibody
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
M2392
Meprobamate, analytical standard
C9H18N2O4
M0400000
Meprobamate, European Pharmacopoeia (EP) Reference Standard
C9H18N2O4
SAB2101427
Anti-MAP2K3 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101428
Anti-MAP2K3 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB4300391
Anti-MAP2K3 (Ab-189) antibody produced in rabbit, affinity isolated antibody
SAB4504622
Anti-phospho-MAP2K3 (pThr222) antibody produced in rabbit, affinity isolated antibody