EMAIL THIS PAGE TO A FRIEND
Food Chemistry
An improved UHPLC-UV method for separation and quantification of carotenoids in vegetable crops.
PMID 25038701
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7906
1-Butanol, for molecular biology, ≥99%
C4H10O
19422
1-Butanol, analytical standard
C4H10O
281549
1-Butanol, anhydrous, 99.8%
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
19420
1-Butanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
270679
1-Butanol, for HPLC, 99.8%
C4H10O
442665
1-Butanol, analytical standard
C4H10O
03-4585
1-Butanol, for HPLC
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
B1378
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
C15H24O
34750
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
C15H24O
03762
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph.Eur.
C15H24O
40395
α-Carotene, ≥95.0% (HPLC)
C40H56
W217816
Butyl alcohol, natural, ≥99.5%, FCC, FG
C4H10O
W217808
Butyl alcohol, ≥99.9%, FCC
C4H10O
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O
W218405
Butylated hydroxytoluene, ≥99%, FCC, FG
C15H24O
PHR1117
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
C15H24O
07168
Lutein, analytical standard
C40H56O2
75051
Lycopene, analytical standard
C40H56
L9879
Lycopene, ≥90%, from tomato
C40H56
36275
Lycopene, Redivivo, 10% FS, ~10% in corn oil, ≥95.0% (sum of isomers)
C40H56
X6250
Xanthophyll, from marigold
C40H56O2
14681
Zeaxanthin, analytical standard
C40H56O2