EMAIL THIS PAGE TO A FRIEND
Food Chemistry
Stability and biological activity of wild blueberry (Vaccinium angustifolium) polyphenols during simulated in vitro gastrointestinal digestion.
PMID 25038707
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4500
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
C17H20N4O6
R7649
(−)-Riboflavin, meets USP testing specifications
C17H20N4O6
R0508
(−)-Riboflavin, acrylamide photopolymerization tested
C17H20N4O6
R9504
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C17H20N4O6
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
64340
DL-Methionine, ≥99.0% (NT)
C5H11NO2S
M0950000
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
C5H11NO2S
G7384
Gallic acid, 97.5-102.5% (titration)
C7H6O5
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
51280
Hemin, BioXtra, from Porcine, ≥97.0% (HPLC)
C34H32ClFeN4O4
H9039
Hemin, from bovine, ≥90%
C34H32ClFeN4O4
W326305
L-Cysteine, ≥97%, FG
C3H7NO2S
168149
L-Cysteine, 97%
C3H7NO2S
C7352
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
C3H7NO2S
C5360
L-Cysteine, PharmaGrade, from non-animal source, Ajinomoto, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S
30089
L-Cysteine, BioUltra, ≥98.5% (RT)
C3H7NO2S
778672
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
C3H7NO2S
95437
L-Cysteine, certified reference material, TraceCERT®
C3H7NO2S
M5625
Menadione, crystalline
C11H8O2
M9429
Menadione, meets USP testing specifications
C11H8O2
1381006
Menadione, United States Pharmacopeia (USP) Reference Standard
C11H8O2
M0300000
Menadione, European Pharmacopoeia (EP) Reference Standard
C11H8O2
M57405
Menadione, 98%
C11H8O2
PHR1411
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
C11H8O2
47775
Menadione (K3), analytical standard
C11H8O2
M0960000
Methionine, European Pharmacopoeia (EP) Reference Standard
C5H11NO2S
PHR1054
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N4O6
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
R0600000
Riboflavin, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6