EMAIL THIS PAGE TO A FRIEND
Journal of Paediatrics and Child Health
High flow nasal cannula for continuous positive airway pressure weaning in preterm neonates: A single-centre experience.
PMID 25039700