EMAIL THIS PAGE TO A FRIEND
Transfusion
AS-7 improved in vitro quality of red blood cells prepared from whole blood held overnight at room temperature.
PMID 25039791
Abstract
Extended room temperature (RT) hold of whole blood (WB) may affect the quality of red blood cell (RBC) components produced from these donations. The availability of better RBC additive solutions (ASs) may help reduce the effects. A new AS, AS-7 (SOLX, Haemonetics Corporation), was investigated for improved in vitro quality of RBCs prepared from WB held overnight at RT. Sixteen WB units were held for 21.4 hours ± 40 minutes at 22°C on cooling plates before processing. Each pair of ABO-matched WB units were pooled, divided into a WB filter pack containing saline-adenine-glucose-mannitol (control) and a LEUKOSEP WB-filter pack containing SOLX, and processed according to manufacturer's instructions. RBCs were stored at 2 to 6°C and sampled weekly until expiry. Glycophorin A (GPA+) and annexin V-binding microparticles (MPs) were quantitated using flow cytometry. Osmotic fragility, intracellular pH (pHi), adenosine triphosphate (ATP), 2,3-diphosphoglycerate (2,3-DPG), and routine quality variables were measured. Adhesion of RBCs to human endothelial cells (ECs) was evaluated by flow perfusion under low shear stress (0.5 dyne/cm(2) ), similar to low blood flow in microvessels. ATP and 2,3-DPG levels were improved for SOLX-RBCs. SOLX-RBCs maintained higher pHi, increased resistance to hypotonic stress, and reduced numbers of GPA+ MPs. No significant difference was observed between annexin V binding to MPs or adhesion of RBCs to ECs under shear stress. SOLX-stored RBCs showed increased osmotic resistance, pHi, and reduced GPA+ MPs and together with higher ATP and 2,3-DPG levels demonstrated improved in vitro RBC quality measures during 42 days of storage.