EMAIL THIS PAGE TO A FRIEND
British Journal of Pharmacology
Glutathione administration reduces mitochondrial damage and shifts cell death from necrosis to apoptosis in ageing diabetic mice hearts during exercise.
PMID 25039894