EMAIL THIS PAGE TO A FRIEND
Cancer
Coronary artery disease detected by coronary computed tomography angiography in adult survivors of childhood Hodgkin lymphoma.
PMID 25041978
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57660
Iodine, purum p.a., ≥99.5% (RT)
I2