EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Recognitive nano-thin-film composite beads for the enantiomeric resolution of the metastatic breast cancer drug aminoglutethimide.
PMID 25042439