EMAIL THIS PAGE TO A FRIEND
International journal of obesity (2005)
Gamma-tocotrienol attenuates high-fat diet-induced obesity and insulin resistance by inhibiting adipose inflammation and M1 macrophage recruitment.
PMID 25042861
Abstract
We have previously demonstrated that gamma tocotrienol (γT3) potently inhibits adipocyte hyperplasia in human adipose-derived stem cells (hASCs). In this study, our objective was to investigate the γT3 effects on early-onset obesity, inflammation and insulin resistance in vivo. Young C57BL/6J mice were fed a high-fat (HF) diet supplemented with 0.05% γT3 for 4 weeks. The concentrations of γT3 in plasma and adipose tissue were measured using high-performance liquid chromatography. Effects of γT3 on body weight gain, adipose volume, plasma levels of fasting glucose, insulin (enzyme-linked immunosorbent assay (ELISA)), proinflammatory cytokines (mouse cytokine array), insulin signaling (western blotting) and gene expression (quantitative real-time PCR, qPCR) in the liver and adipose tissue were examined. Influences of γT3 on [3H]-2-deoxyglucose uptake and lipopolysaccharide (LPS)-mediated NFκB signaling (western blotting) were assessed in hASCs. Effects of γT3 on macrophage M1/M2 activation were investigated using qPCR in mouse bone marrow-derived macrophages. After a 4-week treatment, γT3 accumulated in adipose tissue and reduced HF diet-induced weight gain in epididymal fat, mesenteric fat and the liver. Compared with HF diet-fed mice, HF+γT3-fed mice were associated with (1) decreased plasma levels of fasting glucose, insulin and proinflammatory cytokines, (2) improved glucose tolerance and (3) enhanced insulin signaling in adipose tissue. There were substantial decreases in macrophage specific markers, and monocyte chemoattractant protein-1, indicating that γT3 reduced the recruitment of adipose tissue macrophages (ATMs). In addition, γT3 treatment in human adipocytes resulted in (1) activation of insulin-stimulated glucose uptake and (2) a significant suppression of MAP kinase and NFκB activation. In parallel, γT3 treatment led to a reduction of LPS-mediated M1 macrophage polarization. Our results demonstrated that γT3 ameliorates HF diet-mediated obesity and insulin resistance by inhibiting systemic and adipose inflammation, as well as ATM recruitment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3179
2-Deoxy-D-glucose, BioXtra, ≥98%
C6H12O5