EMAIL THIS PAGE TO A FRIEND
The Journal of Comparative Neurology
Characterization of axons expressing the artemin receptor in the female rat urinary bladder: a comparison with other major neuronal populations.
PMID 25043933