EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Sodium vanadium oxide: a new material for high-performance symmetric sodium-ion batteries.
PMID 25044526
Abstract
Room-temperature sodium-ion batteries have the potential to become the technology of choice for large-scale electrochemical energy storage because of the high sodium abundance and low costs. However, not many materials meet the performance requirements for practical applications. Here, we report a novel sodium-ion battery electrode material, Na(2.55)V(6)O(16)⋅0.6 H(2)O, that shows significant capacities and stabilities at high current rates up to 800 mA g(-1). X-ray photoelectron spectroscopy measurements are carried out to better understand the underlying reactions. Moreover, due to the different oxidation states of vanadium, this material can also be employed in a symmetric full cell, which would decrease production costs even further. For these full cells, capacity and stability tests are conducted using various cathode:anode mass ratios.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357162
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
V
357170
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
V
266205
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
V
262935
Vanadium, powder, −325 mesh, 99.5% trace metals basis
V
266175
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
V
262927
Vanadium, turnings, 99.7% trace metals basis
V
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V
GF26068855
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
V
GF67217430
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
V
GF78288946
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
V
GF08542975
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
V
GF58606409
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
V
GF02179672
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
V