EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International Edition in English)
Organic chemistry as a language and the implications of chemical linguistics for structural and retrosynthetic analyses.
PMID 25044611