EMAIL THIS PAGE TO A FRIEND
Journal of Separation Science
Preparation of Keggin-type phosphomolybdate by a one-step solid-state reaction at room temperature and its application in protein adsorption.
PMID 25044716