EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
A derivatization and validation strategy for determining the spatial localization of endogenous amine metabolites in tissues using MALDI imaging mass spectrometry.
PMID 25044893
Abstract
Imaging mass spectrometry (IMS) studies increasingly focus on endogenous small molecular weight metabolites and consequently bring special analytical challenges. Since analytical tissue blanks do not exist for endogenous metabolites, careful consideration must be given to confirm molecular identity. Here, we present approaches for the improvement in detection of endogenous amine metabolites such as amino acids and neurotransmitters in tissues through chemical derivatization and matrix-assisted laser desorption/ionization (MALDI) IMS. Chemical derivatization with 4-hydroxy-3-methoxycinnamaldehyde (CA) was used to improve sensitivity and specificity. CA was applied to the tissue via MALDI sample targets precoated with a mixture of derivatization reagent and ferulic acid as a MALDI matrix. Spatial distributions of chemically derivatized endogenous metabolites in tissue were determined by high-mass resolution and MS(n) IMS. We highlight an analytical strategy for metabolite validation whereby tissue extracts are analyzed by high-performance liquid chromatography (HPLC)-MS/MS to unambiguously identify metabolites and distinguish them from isobaric compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29208
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide, for protein sequence analysis (by MALDI-MS), ≥98.5%
C33H39BrNO13P
39068
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride, ≥97.5% (AT)
C14H14ClN3O2S
382051
4-Hydroxy-3-methoxycinnamaldehyde, 98%
C10H10O3
W228613
Cinnamaldehyde, natural, ≥95%, FG
C9H8O
502219
Dabsyl chloride, for HPLC derivatization
C14H14ClN3O2S
D2625
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
C12H12ClNO2S
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S
03641
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
C12H12ClNO2S
Y0001013
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
C10H10O4
G0355000
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
C5H9NO4
1380105
Melatonin, United States Pharmacopeia (USP) Reference Standard
C13H16N2O2
C80687
trans-Cinnamaldehyde, 99%
C9H8O
239968
trans-Cinnamaldehyde, ≥99%
C9H8O
W228605
trans-Cinnamaldehyde, FCC, FG
C9H8O
06536
trans-Cinnamaldehyde, analytical standard
C9H8O
128708
trans-Ferulic acid, 99%
C10H10O4
W518301
trans-Ferulic acid, ≥99%
C10H10O4
46278
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C10H10O4
52229
trans-Ferulic acid, certified reference material, TraceCERT®
C10H10O4
T2610000
Tryptophan, European Pharmacopoeia (EP) Reference Standard
C11H12N2O2