EMAIL THIS PAGE TO A FRIEND
BioMed Research International
An in vivo microdialysis study of FLZ penetration through the blood-brain barrier in normal and 6-hydroxydopamine induced Parkinson's disease model rats.
PMID 25045708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8523
6-Hydroxydopamine hydrobromide, ≥90% (HPLC), solid
C8H11NO3 · HBr
H116
6-Hydroxydopamine hydrobromide, contains ascorbic acid as stabilizer, ≥98% (HPLC)
C8H11NO3 · HBr
162957
6-Hydroxydopamine hydrobromide, 95%
C8H11NO3 · HBr
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid
C6H8O6
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
95212
L-Ascorbic acid, tested according to Ph.Eur.
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa