EMAIL THIS PAGE TO A FRIEND
Chemosphere
Comparison of TiO2 nanoparticle and graphene-TiO2 nanoparticle composite phototoxicity to Daphnia magna and Oryzias latipes.
PMID 25048889
Abstract
With a dramatic rise in complexity, needs of nanotoxicology research go beyond simple forms of nanomaterials. This study compared the phototoxicity of nano-TiO2 and graphene-TiO2 nanocomposite (GNP). GNP was synthesized based on a hydrothermal method, which simultaneously performed the reduction of graphene oxide and nano-TiO2 loading. A series of acute toxicity tests of nano-TiO2, graphene and GNP was performed on two aquatic organisms, Daphnia magna and Oryzias latipes. Fast and substantial agglomeration and sedimentation of nanoparticles in test media and surface attachment of nano-TiO2 and GNP on D. magna surface was observed. Similar phototoxicity of nano-TiO2 and GNP for both species existed, though compared with nano-TiO2, GNP had a 2.3-fold increase in visible light photocatalytic ROS generation. In summary, this study demonstrated the significance of illumination spectrum, particle behavior, and species sensitivity on nanophototoxicity, and the needs for research on increasingly sophisticated functional materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282863
Graphite, powder, <20 μm, synthetic
C
496596
Graphite, powder, <45 μm, ≥99.99% trace metals basis
C
332461
Graphite, flakes
C
496588
Graphite, powder, <150 μm, 99.99% trace metals basis
C
496553
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
C
496537
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
C
GF03980932
Titanium, mesh, 150x150mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF16031843
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF71783354
Titanium, wire reel, 100m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF76735963
Titanium, wire reel, 10m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF29374005
Titanium, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Ti
GF34739837
Titanium, sphere, 200pcs, diameter 3mm, 99.6+%
Ti
GF64061760
Titanium, rod, 1000mm, diameter 16mm, annealed, 99.6+%
Ti
GF26694771
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Ti
GF70333376
Titanium, wire reel, 10m, diameter 0.05mm, hard, 99.8%
Ti
GF96834493
Titanium, rod, 1000mm, diameter 5mm, annealed, 99.6+%
Ti
GF16213553
Titanium, wire reel, 10m, diameter 0.125mm, annealed, 99.6+%
Ti
GF76266423
Titanium, wire reel, 0.5m, diameter 1.0mm, annealed, 99.6+%
Ti
GF46132534
Titanium, wire reel, 100m, diameter 0.25mm, as drawn, 99.6+%
Ti
GF76772391
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Ti
GF98611623
Titanium, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Ti
GF66991179
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Ti
GF58567067
Titanium, rod, 100mm, diameter 5mm, annealed, 99.6+%
Ti
GF33043069
Titanium, rod, 100mm, diameter 30mm, annealed, 99.6+%
Ti
GF34996685
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Ti
GF13900205
Titanium, tube, 100mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Ti
GF71724124
Titanium, tube, 500mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Ti
GF50643479
Titanium, wire reel, 5m, diameter 1.5mm, as drawn, 99.6+%
Ti
GF90688449
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF51690663
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Ti