EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Parallel ultra high pressure liquid chromatography-mass spectrometry for the quantification of HIV protease inhibitors using dried spot sample collection format.
PMID 25049214
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0685 Amprenavir, ≥98% (HPLC)
C25H35N3O6S
SML0937
Darunavir, ≥98% (HPLC)
C27H37N3O7S
PZ0013
Nelfinavir mesylate hydrate, ≥98% (HPLC)
C32H45N3O4S · CH3SO3H · xH2O